Skip to Content
Merck
Histamine bisphosphate monohydrate

Histamine bisphosphate monohydrate

Histamine bisphosphate monohydrate
Synonym(s):
2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(4-Imidazolyl)ethylamine diphosphate salt monohydrate, Histamine diphosphate salt monohydrate
Empirical Formula (Hill Notation):
C5H9N3 · 2H3PO4 · H2O
CAS Number:
Molecular Weight:
325.15
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing