Skip to Content
Merck
All Photos(3)

Key Documents

H4002

Sigma-Aldrich

DL-3-Hydroxynorvaline

≥98% (TLC)

Synonym(s):

HNV, α-Amino-β-hydroxyvaleric acid, 2-Amino-3-hydroxypentanoic acid, 3-Hydroxy-2-aminopentanoic acid, DL-β-Hydroxynorvaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO3
CAS Number:
Molecular Weight:
133.15
Beilstein:
1722350
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

DL-3-Hydroxynorvaline, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

CCC(O)C(N)C(O)=O

InChI

1S/C5H11NO3/c1-2-3(7)4(6)5(8)9/h3-4,7H,2,6H2,1H3,(H,8,9)

InChI key

LGVJIYCMHMKTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

3-Hydroxynorvaline (HNV) is a microbial, α amino acid threonine analogue which has antiviral activity (herpes virus) and is toxic to mammalian cells (embryotoxic and teratogenic) presumably via incorporation into proteins. 3-Hydroxynorvaline may be used as an unnatural amino acid to study the fidelity of protein translation at the level of acyl-tRNA editing. 3-Hydroxynorvaline may also be used to study enzymes and molecules involved in threonine metabolism.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F A Barile et al.
Archives of biochemistry and biophysics, 270(1), 294-301 (1989-04-01)
Amino acid substitutions in collagen that impair folding of the triple helix result in significant increases in intracellular degradation of newly synthesized collagen. We have studied the effects of agents that cause other kinds of defects in collagen: hydroxynorvaline, a
Y J Gordon et al.
Current eye research, 5(2), 93-99 (1986-02-01)
The present study examined the anti-herpetic effect of the glycoprotein inhibitors, hydroxynorvaline and 2-deoxyglucose, alone and in combination with trifluridine on murine ocular herpes. Following ocular inoculation with a large dose of HSV-1 RE strain (10(6) pfu), ICR mice were
P A Docherty et al.
The Journal of biological chemistry, 260(19), 10847-10855 (1985-09-05)
The threonine analog beta-hydroxynorvaline (Hnv) is an inhibitor of asparagine-linked glycosylation. In the presence of the analog hepatocytes synthesized immunoreactive alpha 1-acid glycoprotein with 0-6 oligosaccharide chains. Pulse-chase experiments were conducted to compare the rates of secretion of alpha 1-acid
C M Alarcón et al.
Molecular and cellular biology, 17(10), 5968-5975 (1997-10-07)
The peptidyl-prolyl isomerase FKBP12 was originally identified as the intracellular receptor for the immunosuppressive drugs FK506 (tacrolimus) and rapamycin (sirolimus). Although peptidyl-prolyl isomerases have been implicated in catalyzing protein folding, the cellular functions of FKBP12 in Saccharomyces cerevisiae and other
E G Green et al.
The Journal of steroid biochemistry and molecular biology, 40(1-3), 421-429 (1991-01-01)
We have investigated the ability of amino acid analogues of serine and threonine to inhibit the increase in steroidogenesis elicited by addition of ACTH or cAMP in cells isolated from the rat adrenal cortex. We have found that the serine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service