Skip to Content
Merck
All Photos(1)

Key Documents

H3631

Sigma-Aldrich

Hyaluronidase from bovine testes

Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid

Synonym(s):

Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine testis

Quality Level

conjugate

conjugate (Glucosaminoglycan)

type

Type VI-S

form

lyophilized powder

specific activity

3,000-15,000 units/mg solid

mol wt

~55 kDa (four subunits of 14 kDa each)

UniProt accession no.

relevant disease(s)

cancer

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hyaluronidase degrades hyaluronan and has been found to be inappropriately regulated during cancer progression.
These enzymes randomly cleave β-N-acetylhexosamine-[1→4] glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.

Unit Definition

One unit will degrade 0.75 μg of the polysaccharide hyaluronic acid per minute at pH 5.35 at 37 ºC (as measured by turbidimetric absorbance (λ: 600nm) when complexed with BSA after 45 minutes).

Preparation Note

Chromatographically purified

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tongcui Ma et al.
eLife, 11 (2022-07-06)
High-parameter single-cell phenotyping has enabled in-depth classification and interrogation of immune cells, but to date has not allowed for glycan characterization. Here, we develop CyTOF-Lec as an approach to simultaneously characterize many protein and glycan features of human immune cells
Dong-Ru Ho et al.
International journal of urology : official journal of the Japanese Urological Association, 18(7), 525-531 (2011-05-25)
To investigate how hyaluronic acid (HA) affects nerve growth factor (NGF) production and bladder overactivity in a cyclophosphamide (CYP)-induced cystitis rat model. Female Sprague-Dawley rats received three intermittent intraperitoneal injections of CYP (75 mg/kg) or saline. Before or after CYP injection
Tongcui Ma et al.
eLife, 9 (2020-05-27)
The female reproductive tract (FRT) is the most common site of infection during HIV transmission to women, but viral remodeling complicates characterization of cells targeted for infection. Here, we report extensive phenotypic analyses of HIV-infected endometrial cells by CyTOF, and
Sialic acids rather than glycosaminoglycans affect normal and sickle red blood cell rheology by binding to four major sites on fibrinogen.
Frank Gondelaud et al.
American journal of hematology, 95(4), E77-E80 (2020-01-07)
Shared decision making, thrombotic thrombocytopenic purpura, and caplacizumab.
Senthil Sukumar et al.
American journal of hematology, 95(4), E76-E77 (2020-01-03)

Articles

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

See All

Protocols

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

Questions

  1. We bought this product. H3631, Batch Number: SLCH2685. Could you please provide the info on componants like, what are other componants present along with Hyaluronidase which is 3909/mg of solid (40ug, what is the remaining 940ug)?

    1 answer
    1. This product is a further purification of product H3506 and may contain some residual salts, such as sodium chloride.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service