H1115000
Hyaluronidase
European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
pharmaceutical primary standard
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
InChI
1S/C18H14O7/c19-13(11-2-4-15-17(6-11)25-9-23-15)7-12(18(20)21)10-1-3-14-16(5-10)24-8-22-14/h1-6,12H,7-9H2,(H,20,21)
InChI key
PPEBBOHQEAQKSW-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Hyaluronidase EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Packaging
The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.
Other Notes
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nature communications, 9(1), 2574-2574 (2018-07-04)
Metabolic reprogramming greatly contributes to the regulation of macrophage activation. However, the mechanism of lipid accumulation and the corresponding function in tumor-associated macrophages (TAMs) remain unclear. With primary investigation in colon cancer and confirmation in other cancer models, here we
Cancer biology & therapy, 20(7), 967-978 (2019-03-22)
Nontumour cells in the tumour microenvironment, especially fibroblasts, contribute to tumour progression and metastasis. The occurrence and evolution of colorectal cancer (CRC) is closely related to cancer-associated fibroblasts (CAFs). The aim of this work was to evaluate the effects of
Asian journal of neurosurgery, 13(4), 1101-1107 (2018-11-22)
Spinal cord injury (SCI) as one of the most important diseases of central nervous system (CNS) without any definite treatment is still growing in incidence. In addition to trauma, some surgeries such as cardiac and thoracic aorta surgery may result
Erratum to: Isolation and Characterization of Aortic Dendritic Cells and Lymphocytes in Atherosclerosis.
Methods in molecular biology (Clifton, N.J.), 1559, E1-E1 (2017-10-19)
Turkish journal of urology, 45(3), 164-170 (2019-03-01)
Studies showed a decrease of the semen analysis parameters and an increase in the average age of first-time fathers over the past several decades. The aim of the present study was to assess the influence of paternal age on semen
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service