Skip to Content
Merck
All Photos(1)

Key Documents

H102

Supelco

Haloperidol metabolite II

analytical standard

Synonym(s):

(±)-4-(4-Chlorophenyl)-α-(4-fluorophenyl)-4-hydroxy-1-piperidinebutanol, Reduced Haloperidol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClFNO2
CAS Number:
Molecular Weight:
377.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

solubility

DMSO: 10 mg/mL

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

OC(CCCN1CCC(O)(CC1)c2ccc(Cl)cc2)c3ccc(F)cc3

InChI

1S/C21H25ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,20,25-26H,1-2,11-15H2

InChI key

WNZBBTJFOIOEMP-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Lee et al.
Psychiatry and clinical neurosciences, 52 Suppl, S193-S198 (1999-01-23)
Along with recent increased interest in the selective serotonin reuptake inhibitors, a number of studies has been undertaken to observe interactions with different drugs. When selective serotonin reuptake inhibitor was administered together with antipsychotics to schizophrenics showing depressive or obsessive
L Pan et al.
European journal of clinical pharmacology, 55(8), 599-604 (1999-12-14)
To investigate in vitro which CYP isoforms (CYP1A2, CYP2D6 and CYP3A4) are involved in the biotransformation of haloperidol (HAL) and reduced haloperidol (RHAL). The biotransformation of HAL and RHAL is evaluated by measuring HAL and RHAL remaining after incubation with
D W Eyles et al.
Psychopharmacology, 116(2), 161-166 (1994-10-01)
In patients taking haloperidol (HP), circulating concentrations of reduced haloperidol (RHP increase disproportionately to the dose or concentration of the parent drug. In the current study, we tested the hypothesis that the nonlinearity is due to preferential saturation of the
M Aravagiri et al.
Journal of chromatography. B, Biomedical applications, 656(2), 373-381 (1994-06-17)
A simple and highly sensitive liquid chromatographic method with electrochemical detection for the simultaneous determination of haloperidol and its metabolite reduced haloperidol in human plasma has been developed. The sample preparation for the analysis involves a simple one-step extraction procedure
S Ulrich et al.
Journal of chromatography. B, Biomedical applications, 663(2), 289-296 (1995-01-20)
A gas chromatographic method using a HP-5 megabore capillary and nitrogen-phosphorus selective detection for the quantitative analysis of haloperidol (H) and reduced haloperidol (RH) in human serum or plasma is described. A 3-step liquid-liquid extraction is applied. The extraction yield

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service