Skip to Content
Merck
All Photos(1)

Key Documents

H139

Sigma-Aldrich

HA-1077 dihydrochloride

solid, ≥98% (HPLC)

Synonym(s):

1-(5-Isoquinolinylsulfonyl)homopiperazine dihydrochloride, Fasudil dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N3O2S · 2HCl
CAS Number:
Molecular Weight:
364.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

H2O: >10 mg/mL

SMILES string

Cl[H].Cl[H].O=S(=O)(N1CCCNCC1)c2cccc3cnccc23

InChI

1S/C14H17N3O2S.2ClH/c18-20(19,17-9-2-6-15-8-10-17)14-4-1-3-12-11-16-7-5-13(12)14;;/h1,3-5,7,11,15H,2,6,8-10H2;2*1H

InChI key

NOXXIYDYFSNHDF-UHFFFAOYSA-N

Application

HA-1077 dihydrochloride has been used:
  • to relax cantilevers
  • in the functional assessment of isolated coronary arteries
  • to investigate the effect of azaindole-1, fasudil on acute hypoxic pulmonary vasoconstriction (HPV)

Biochem/physiol Actions

HA-1077/fasudil, is a Rho kinase inhibitor. This ROCK inhibitor has the ability to increase cognitive deficits in animals, that have Alzheimer′s disease (AD). In humans, fasudil possesses high therapeutic effects against cerebral vasospasm after subarachnoid hemorrhage (SAH). HA-1077 also acts as an inhibitor of protein kinases, like myosin light chain kinase and Rho associated kinase. Hence, it can block myosin phosphorylation.
Novel vasodilator agent which inhibits vascular smooth muscle contraction by acting as an intracellular Ca2+ antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fasudil exerts cardioprotection in Animal Models of Myocardial Ischemia/Reperfusion injury: A Meta-analysis and Review of Preclinical Evidence and Possible Mechanisms
Huang Y, et al.
Frontiers in Pharmacology, 9, 1083-1083 (2018)
M Seto et al.
European journal of pharmacology, 195(2), 267-272 (1991-03-26)
We examined the effects of HA1077, a potent inhibitor of protein kinases in vitro, on the relationship between tension and myosin-light chain (MLC20) phosphorylation in the initial phase of contraction of the rabbit aorta. The dose-response curve of HA1077 for
Therapeutic efficacy of azaindole-1 in experimental pulmonary hypertension
Dahal B K, et al.
European heart journal. Cardiovascular pharmacotherapy, erj01403-erj02009 (2010)
The kinase inhibitor fasudil (HA-1077) reduces intimal hyperplasia through inhibiting migration and enhancing cell loss of vascular smooth muscle cells
Negoro N, et al.
Biochemical and Biophysical Research Communications, 262(1), 211-215 (1999)
Acute pergolide exposure stiffens engineered valve interstitial cell tissues and reduces contractility in vitro
Capulli A K, et al.
Cardiovascular Pathology : the Official Journal of the Society For Cardiovascular Pathology, 25(4), 316-324 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service