Recommended Products
biological source
mouse thymus
Quality Level
description
Mouse DBA/2 lymphoma L5178Y tk+/-
growth mode
Suspension
morphology
Small rounded cells
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
1 of 4
This Item | H-129 | L-901 | T-911 |
---|---|---|---|
single component solution | single component solution | single component solution | single component solution |
liquid | liquid | liquid | liquid |
300 | 300 | - | 300 |
certified reference material | certified reference material | certified reference material | certified reference material |
2-8°C | −20°C | −20°C | 2-8°C |
Cell Line Description
Culture Medium
Subculture Routine
Other Notes
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service