Skip to Content
Merck
All Photos(1)

Key Documents

GW22511

Sigma-Aldrich

Anti-HA tag antibody produced in chicken

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

manufacturer/tradename

Genway 15-288-22511

technique(s)

indirect ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

The HA tag is derived from an epitope of the influenza hemagglutinin protein, which has been extensively used as a general fusion tag in expression vectors. It has antigenic sites, cleavage sites, receptor binding sites, and fusion peptides mediating membrane fusion.

Immunogen

Immunogen Sequence: GI # 1373274, sequence 143-151
Synthetic peptide sequence of HA tag.

Application

Anti-HA tag antibody produced in chicken has been used in immunostaining (1:100).

Biochem/physiol Actions

The hemagglutinin (HA) protein is a key factor of influenza A virus pathogenicity.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eva Böttcher-Friebertshäuser et al.
Current topics in microbiology and immunology, 385, 3-34 (2014-07-18)
The hemagglutinin (HA) is a prime determinant of the pathogenicity of influenza A viruses. It initiates infection by binding to cell surface receptors and by inducing membrane fusion. The fusion capacity of HA depends on cleavage activation by host proteases
Nongluk Sriwilaijaroen et al.
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 88(6), 226-249 (2012-06-26)
Influenza virus hemagglutinin (HA) contains antigenic sites recognized by the host immune system, cleavage sites cleaved by host proteases, receptor binding sites attaching to sialyl receptors on the target cell, and fusion peptides mediating membrane fusion. Change in an amino
Csaba Bajusz et al.
The FEBS journal, 288(16), 4812-4832 (2021-02-20)
Ezrin-Radixin-Moesin (ERM) proteins play an essential role in the cytoplasm by cross-linking actin filaments with plasma membrane proteins. Research has identified the nuclear localization of ERMs, as well as the involvement of a single Drosophila ERM protein, Moesin, in nuclear
Ildikó Kristó et al.
Biochimica et biophysica acta. Molecular cell research, 1864(10), 1589-1604 (2017-05-31)
Current models imply that the evolutionarily conserved, actin-binding Ezrin-Radixin-Moesin (ERM) proteins perform their activities at the plasma membrane by anchoring membrane proteins to the cortical actin network. Here we show that beside its cytoplasmic functions, the single ERM protein of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service