Skip to Content
Merck
All Photos(3)

Key Documents

BR4760161

BRAND® Titrette® digital bottle-top burette

capacity 50 mL, without interface RS 232

Synonym(s):

Bottle-top Burette, Bottle-top Dispenser, Burette, Dispenser, Titrette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121808
NACRES:
NB.43

feature

without interface RS 232

manufacturer/tradename

BRAND 4760161

volume measuring range

50 mL

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Titrette® is the first bottle-top burette to meet class A error limits for glass burettes. In addition, it stands out for easy titration, a compact design, excellent precision, an exchangeable dosing unit, and an optional PC interface. You can use the Titrette® to titrate quickly and reliably, even with restricted space and independent of available power connections - in the laboratory, in production, or in the field.

Application

Titrating

Features and Benefits

Overview:
  • Simple: Titration within Class A error limits directly from the bottle
  • Easy: Compact and robust
  • Efficient: Fast filling, precise dispensing
  • Separate buttons for on/off, pause, and CLEAR to delete the display and select function
  • Non-slip hand wheels and a smooth-running precision gear set for fast or drip titration make handling reliable.


Limitations on use:
  • Operating temperature from +15 °C to +40 °C (from 59 °F to 104 °F) of instrument and reagent
  • Vapor pressure up to 500 mbar
  • Viscosity up to 500 mm2 /s
  • Altitude: max. 3,000 m above sea level
  • Relative humidity: 20 % to 90%

Components

Items supplied:
  • Titrette bottle-top burette
  • DE-M marking
  • Performance certificate
  • Telescoping filling tube (170 - 330 mm)
  • Recirculation tube
  • 2 micro batteries (AAA/UM4/LR03)
  • 3 bottle adapters PP (GL 45/32, GL 45/S 40 und GL 32/NS 29/32)
  • 2 colored light shield inspection windows


With PC Interface, also includes:
  • Connector cable (Sub-D plug connector, 9-pin)
  • CD with driver software
  • Communication protocol
  • Example application
  • Operating manual

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This article explains cleaning procedures of the BRAND® Titrette® Bottletop Burette.

This article explains cleaning procedures of the BRAND® Titrette® Bottletop Burette.

This article explains cleaning procedures of the BRAND® Titrette® Bottletop Burette.

This article decribes how to use the BRAND® Titrette® Digital Bottletop Burette for quick and reliable titration.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service