Skip to Content
Merck
All Photos(1)

Key Documents

GS90

Sigma-Aldrich

GST T1-1, Recombinant Human

Synonym(s):

glutathione S-transferase theta 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

95% (SDS-PAGE)

form

frozen liquid

specific activity

8.8 units/mg protein

mol wt

25 kDa

concentration

1.95 mg/mL

storage temp.

−70°C

Gene Information

human ... GSTT1(2952)

General description

Glutathione S-transferase theta 1 (GSTT1) and GSTT2/GSTT2B share 55% amino acid sequence identity and may play a role in human carcinogenesis. The GSTT1 gene is haplotype-specific and is absent from 38% of the population. Alternative splicing of this gene results in multiple transcript variants.GSTT1 is an enzyme that in humans is encoded by the GSTT1 gene. The GSTT1 gene contains five exons and four introns and spans 8092bp on genomic DNA. It is mapped to human chromosome 22q11.23. This gene codes for a 240 amino acids, which is mainly expressed in human liver. Based on their biochemical, immunologic, and structural properties, cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: α, κ, μ, ω, π, σ, θ and ζ.
as assayed by the spectrophotometric determination of NADPH oxidation coupled to the glutathione peroxidase activity of GST T1-1 on cumene hydroperoxide (1.5 mM) in the presence of reduced glutathione (1 mM) in 100 mM NaPO4 (pH 7.0) at room temperature.

Biochem/physiol Actions

Glutathione S-transferase theta 1 (GSTT1) is an enzyme that in humans is encoded by the GSTT1 gene. Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Mutation in the gene might lead to the development of lung, head and neck cancer. The GSTs are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.

Storage and Stability

The enzyme should be used by the end-user customer within 1 year of receipt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial glutathione S-transferases: what are they good for?
Vuilleumier, S, et al.
Journal of Bacteriology, 179, 1431-1431 (1997)
Wen Sun et al.
BMC ophthalmology, 15, 84-84 (2015-07-26)
Glutathione S transferase (GST) polymorphisms have been considered as risk factors for age-related cataracts, but the results remain controversial. In this study, we have performed a meta-analysis to evaluate the association between polymorphisms of GSTM1 and GSTT1 and cataract risk.
Ender Coskunpinar et al.
Asian Pacific journal of cancer prevention : APJCP, 15(5), 2319-2321 (2014-04-11)
Our aim was to determine GSTT1 expression levels in left colon tumors and paired normal tissue in order to identify specific alterations in GSTT1 mRNA levels. Alterations in GSTT1 expression in twenty-four left- sided colon tumors and paired cancer free
Dan Wang et al.
Molecular biology reports, 41(11), 7423-7430 (2014-08-05)
Conflicting results in previous case-control studies on the association between Glutathione S-transferase T1 (GSTT1) gene polymorphism and Parkinson's disease (PD) risk have been reported, so we conducted this meta-analysis. We searched and extracted data from 3 Chinese and 3 English
Glutathione and glutathione-dependent enzymes represent a co-ordinately regulated defence against oxidative stress
Hayes, J. D, et al.
Free Radical Research, 31, 273-300 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service