Key Documents
GBL204997
Grace Bio-Labs FlexPlate MP microtiter plate superstructure
size 96 wells, L × W 7.25 mm × 7.25 mm, clear silicone, adhesive one side, with black polystyrene frame
Synonym(s):
incubation chamber cover, silicone microtiter plate cover
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear silicone
L × W:
7.25 mm × 7.25 mm
size:
96 wells
Pricing and availability is not currently available.
material
clear silicone
sterility
non-sterile
feature
lid: no
packaging
pack of 1 ea
manufacturer/tradename
Grace Bio-Labs 204997
L × W
7.25 mm × 7.25 mm
size
96 wells
General description
N/A
Legal Information
FlexPlate MP is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)