BR781732
BRAND® microplate BRANDplates®, immunoGrade
size 96 wells, size 330 μL, flat bottom, black, with transparent bottom
Synonym(s):
BRANDplates®, Immunology, Microplates, high binding, immunoGrade
About This Item
Recommended Products
material
black wells
flat bottom
polystyrene
sterility
non-sterile
feature
lid: no
skirt (F-bottom)
packaging
pack of 5 ea
manufacturer/tradename
BRAND 781732
size
330 μL
96 wells
working volume
330 μL
color
black, with transparent bottom
binding type
high binding surface
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service