Skip to Content
Merck
All Photos(1)

Key Documents

G7772

Sigma-Aldrich

D-Glucose 6-phosphate solution

~1 M in H2O (approx. 260 mg per ml)

Synonym(s):

D(+)-Glucopyranose 6-phosphate, Robison ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O9P
CAS Number:
Molecular Weight:
260.14
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (inorganic)

Quality Level

Assay

≥99% (TLC)

form

liquid

concentration

~1 M in H2O (approx. 260 mg per ml)

technique(s)

thin layer chromatography (TLC): suitable

color

colorless

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(COP(O)(O)=O)C(O)C(O)C(O)C=O

InChI

1S/C6H13O9P/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14/h1,3-6,8-11H,2H2,(H2,12,13,14)

InChI key

VFRROHXSMXFLSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Phosphonate and α-fluorophosphonate analogs of d-glucose 6-phosphate as active-site probes of 1l-myo-inositol 1-phosphate synthase.: This study explores phosphonate and α-fluorophosphonate analogs of d-glucose 6-phosphate, offering insights into their use as probes for understanding enzyme mechanisms, which could impact biochemical research methodologies (Ramos-Figueroa et al., 2023).

Biochem/physiol Actions

D-Glucose 6-phosphate binds and inhibits the activity of sarcoendoplasmic reticulum calcium ATPase in rat brains resulting in a decrease in the accumulation of calcium in the endoplasmic reticulum.

Linkage

Prepared from G 7879.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victor V Lemeshko
Biochimica et biophysica acta, 1838(5), 1362-1371 (2014-01-15)
The simplest mechanism of the generation of the mitochondrial outer membrane potential (OMP) by the VDAC (voltage-dependent anion channel)-hexokinase complex (VHC), suggested earlier, and by the VDAC-glucokinase complex (VGC), was computationally analyzed. Even at less than 4% of VDACs bound
C Kern et al.
Insect molecular biology, 21(4), 456-471 (2012-07-06)
Trehalose phosphate synthase (EC 2.4.1.15; TPS) is the crucial enzyme for the biosynthesis of trehalose, the main haemolymph sugar of insects, and therefore a potential insecticidal molecular target. In this study, we report the functional heterologous expression of Drosophila melanogaster
Gang Zhao et al.
Biomedical chromatography : BMC, 28(7), 947-956 (2013-12-21)
In this study, a simple and sensitive gas chromatography-mass spectrometry method was developed for the study of bioavailability and protein binding and the metabolism of imperatorin in rat. The results showed that the pharmacokinetics of imperatorin after intravenous and oral
Tomoaki Yamamotoya et al.
Biochimica et biophysica acta, 1824(12), 1442-1448 (2012-07-04)
In the studies of Escherichia coli (E. coli), metabolomics analyses have mainly been performed using steady state culture. However, to analyze the dynamic changes in cellular metabolism, we performed a profiling of concentration of metabolites by using batch culture. As
Glucose-6-phosphate reduces calcium accumulation in rat brain endoplasmic reticulum.
Cole, JT.
Frontiers in Neuroscience, 5, 51-51 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service