Skip to Content
Merck
All Photos(1)

Key Documents

G4134

Sigma-Aldrich

Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae)

Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)

Synonym(s):

G-6-P-DH, Zwischenferment

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type IX

Quality Level

form

lyophilized powder

specific activity

200-400 units/mg protein (modified Warburg-Christian)

mol wt

128 kDa

β-NADP and β-NADPH content

≤10 mmol/mol

application(s)

agriculture

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Glucose-6-phosphate dehydrogenase (G6PD) is a key metabolic enzyme of the pentose phosphate pathway. In S. cerevisiae, it is encoded by the ZWF1 gene. G6PD exists as a tetramer in its active form.

Application

Glucose-6-phosphate dehydrogenase is used:
  • To test ketose reductase activity in developing maize endosperm.
  • For recycling microassay of β-NADP and β-NADPH.
  • To measure the intracellular levels of NADPH and total NADP.
  • To measure the nicotinamide adenine dinucleotide (NAD) kinase kinetic assay activity.

Biochem/physiol Actions

Glucose-6-phosphate dehydrogenase catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway.
Glucose-6-phosphate dehydrogenase catalyzes the rate-limiting step in the pentose phosphate pathway. Its function involves the conversion of glucose-6-phosphate to 6-phosphogluconolacetone while generating NADPH, which is essential for the regeneration of glutathione The glutathione system utilizes nicotinamide adenine dinucleotide phosphate hydrogen (NADPH) to effectively eliminate excess hydrogen peroxide. Glucose-6-phosphate dehydrogenase (G6PD) plays an important role in regulating cell growth and survival. Their levels are higher in cells undergoing normal and neoplastic growth. Increased glucose-6-phosphate dehydrogenase activity plays a pivotal role in preventing reactive oxygen species mediated cell death. Glucose-6-phosphate dehydrogenase is over expressed in several cancers whereas its activity is reduced in hyperglycemia. A deficiency in glucose-6-phosphate dehydrogenase causes hemolysis.

Unit Definition

One unit will oxidize 1.0 μmole of D-glucose 6-phosphate to 6-phospho-D-gluconate per min in the presence of NADP at pH 7.4 at 25 °C.

Physical form

Lyophilized powder essentially sulfate-free, containing approx. 20% sodium citrate

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Oh et al.
Molecular and cellular biology, 10(4), 1415-1422 (1990-04-01)
The Saccharomyces cerevisiae GAL5 (PGM2) gene was isolated and shown to encode the major isozyme of phosphoglucomutase. Northern (RNA) blot hybridization revealed that the GAL5 transcript level increased three- to fourfold in response to galactose and was severely repressed in
Revealing the allosterome: systematic identification of metabolite-protein interactions
Orsak T, et al.
Biochemistry, 51(1), 225-232 (2012)
Actin cytoskeleton vulnerability to disulfide stress mediates disulfidptosis
Liu X, et al.
Nature Cell Biology, 25(3), 404-414 (2023)
The global prevalence of glucose-6-phosphate dehydrogenase deficiency: A systematic review and meta-analysis
Nkhoma ET and Poole C
Blood Cells, Molecules and Diseases, 42, 267?278-267?278 (2009)
Impact of glucose-6-phosphate dehydrogenase deficiency on the pathophysiology of cardiovascular disease
Hecker PA, et al.
American Journal of Physiology. Heart and Circulatory Physiology, 304(4), H491-H500 (2013)

Protocols

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service