Skip to Content
Merck
All Photos(3)

Key Documents

G6779

Sigma-Aldrich

Guanine

BioUltra

Synonym(s):

2-Amino-1,7-dihydro-6H-purin-6-one, 2-Amino-6-hydroxypurine, 2-Aminohypoxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5O
CAS Number:
Molecular Weight:
151.13
Beilstein:
9680
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

product line

BioUltra

Assay

≥99% (HPLC)

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

>300 °C (lit.)

solubility

1 M NaOH: 0.1 M, clear, colorless to faintly yellow

cation traces

Al: ≤0.02%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.005%
K: ≤0.005%
Mg: ≤0.005%
Na: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.002%

SMILES string

NC1=Nc2[nH]cnc2C(=O)N1

InChI

1S/C5H5N5O/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)

InChI key

UYTPUPDQBNUYGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kawanishi et al.
Mutation research, 488(1), 65-76 (2001-02-27)
Reactive species generated by chemicals and UV radiation can cause sequence-specific DNA damage and play important roles in mutagenesis, carcinogenesis and aging. We have investigated sequence specificity of oxidative stress-mediated DNA damage by using 32P-labeled DNA fragments obtained from the
Raja Chinnappan et al.
Nucleic acids research, 41(10), e106-e106 (2013-03-26)
Riboswitches are mRNA elements that specifically bind cellular metabolites and control gene expression by modifying their structure. As riboswitches often control essential genes in pathogenic bacteria, riboswitches have been proposed as new targets for antibiotics. High-throughput screening provides a powerful
Alice T Shaw et al.
The New England journal of medicine, 368(25), 2385-2394 (2013-06-04)
In single-group studies, chromosomal rearrangements of the anaplastic lymphoma kinase gene (ALK) have been associated with marked clinical responses to crizotinib, an oral tyrosine kinase inhibitor targeting ALK. Whether crizotinib is superior to standard chemotherapy with respect to efficacy is
Zac Chatterton et al.
Clinical chemistry, 60(7), 995-1003 (2014-05-16)
DNA methylation biomarkers capable of diagnosis and subtyping have been found for many cancers. Fifteen such markers have previously been identified for pediatric acute lymphoblastic leukemia (ALL). Validation of these markers is necessary to assess their clinical utility for molecular
Wieneke A Buikhuisen et al.
The Lancet. Oncology, 14(6), 543-551 (2013-04-16)
Standard chemotherapy does not lead to long-term survival in patients with malignant pleural mesothelioma. Malignant pleural mesothelioma is strongly dependent on vasculature with high vessel counts and high concentrations of serum vascular growth factors. Thalidomide has shown antiangiogenic activity, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service