Skip to Content
Merck
All Photos(1)

Key Documents

G0877

Sigma-Aldrich

Gly-Glu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O5
CAS Number:
Molecular Weight:
204.18
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

Quality Level

form

powder

color

white

mp

155-158 °C

storage temp.

−20°C

SMILES string

NCC(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C7H12N2O5/c8-3-5(10)9-4(7(13)14)1-2-6(11)12/h4H,1-3,8H2,(H,9,10)(H,11,12)(H,13,14)/t4-/m0/s1

InChI key

IEFJWDNGDZAYNZ-BYPYZUCNSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Brand et al.
Journal of cellular physiology, 132(3), 559-564 (1987-09-01)
Pathways of glutamine metabolism in resting and proliferating rat thymocytes were evaluated by in vitro incubations of freshly prepared or 60-h cultured cells for 1-2 h with [U14C]glutamine. Complete recovery of glutamine carbons utilized in products allowed quantification of the
Takayuki K Nemoto et al.
Analytical biochemistry, 548, 78-81 (2018-02-13)
Bacterial dipeptidyl-peptidase (DPP) 7 liberates a dipeptide with a preference for aliphatic and aromatic penultimate residues from the N-terminus. Although synthetic substrates are useful for activity measurements, those currently used are problematic, because they are more efficiently degraded by DPP5.
K Brand et al.
Metabolism: clinical and experimental, 38(8 Suppl 1), 29-33 (1989-08-01)
Pathways of glutamine metabolism in resting and proliferating rat thymocytes and established human T- and B-lymphoblastoid cell lines were evaluated by in vitro incubations of freshly prepared or cultured cells for one to two hours with [U14C]glutamine. Complete recovery of
C Winckler et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 169(7), 495-500 (1999-12-14)
Characteristics of dipeptide transport in pig jejunum were investigated in vitro by applying the Ussing-chamber technique and mucosal uptake studies. Addition of both glycyl-L-glutamine and glycyl-L-sarcosine (20 mmol.l-1) to the mucosal buffer solution significantly increased the short-circuit current by 2.60
Intravenous use of glutamine in peptide form: clinical applications of old and new observations.
S A Adibi
Metabolism: clinical and experimental, 38(8 Suppl 1), 89-92 (1989-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service