Skip to Content
Merck
All Photos(1)

Key Documents

G3127

Sigma-Aldrich

Gly-Sar

≥98% (TLC)

Synonym(s):

Glycyl-sarcosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CON(CH3)CH2COOH
CAS Number:
Molecular Weight:
146.14
Beilstein:
1768450
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Gly-Sar,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white

mp

198 °C

storage temp.

−20°C

SMILES string

CN(CC(O)=O)C(=O)CN

InChI

1S/C5H10N2O3/c1-7(3-5(9)10)4(8)2-6/h2-3,6H2,1H3,(H,9,10)

InChI key

VYAMLSCELQQRAE-UHFFFAOYSA-N

Gene Information

human ... SLC15A1(6564)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hovhannes J Gukasyan et al.
Pharmaceutical research, 34(12), 2488-2497 (2017-08-24)
Studies were conducted in primary cultured rat alveolar epithelial cell monolayers to characterize peptide transporter expression and function. Freshly isolated rat lung alveolar epithelial cells were purified and cultured on permeable support with and without keratinocyte growth factor (KGF). Messenger
Jinling Wang et al.
International journal of nanomedicine, 13, 7997-8012 (2018-12-13)
Polymeric micelles (PMs) hold promise for improving solubility and oral absorption of poorly soluble drugs. Unfortunately, the oral absorption of PMs is also limited by intestinal epithelium. To improve the oral delivery efficiency of micelles, transporter-mediated micelles could enhance the
Junji Miyabe et al.
Journal of pharmacological sciences, 139(3), 215-222 (2019-03-06)
Boron neutron capture therapy (BNCT) is a radiotherapy utilizing the neutron capture and nuclear fission reaction of 10B taken up into tumor cells. The most commonly used boron agent in BNCT, p-borono-l-phenylalanine (BPA), is accumulated in tumors by amino acid
Ina Hubatsch et al.
Journal of medicinal chemistry, 50(21), 5238-5242 (2007-09-25)
The hPepT1-mediated transport properties of a series of 11 synthesized beta- and gamma-peptides have been studied in Caco-2 cells. The results show that several of the compounds interact with the peptide transporter, but only two beta-dipeptides act as substrates and
Ana S Gomes et al.
American journal of physiology. Cell physiology, 318(1), C191-C204 (2019-10-31)
Peptide transporter 1 (PepT1) mediates the uptake of dietary di-/tripeptides in vertebrates. However, in teleost fish gut, more than one PepT1-type transporter might operate, because of teleost-specific whole gen(om)e duplication event(s) that occurred during evolution. Here, we describe a novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service