Skip to Content
Merck
All Photos(2)

Key Documents

G0424

Sigma-Aldrich

Corilagin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H22O18
CAS Number:
Molecular Weight:
634.45
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

Quality Level

form

powder

solubility

H2O: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@H]2COC(=O)c3cc(O)c(O)c(O)c3-c4c(O)c(O)c(O)cc4C(=O)O[C@@H]1[C@@H](O)[C@@H](O2)OC(=O)c5cc(O)c(O)c(O)c5

InChI

1S/C27H22O18/c28-9-1-6(2-10(29)16(9)32)24(39)45-27-22(38)23-19(35)13(43-27)5-42-25(40)7-3-11(30)17(33)20(36)14(7)15-8(26(41)44-23)4-12(31)18(34)21(15)37/h1-4,13,19,22-23,27-38H,5H2/t13-,19-,22-,23+,27+/m1/s1

InChI key

TUSDEZXZIZRFGC-XIGLUPEJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Corilagin is a polyphenolic tannic acid and a major bioactive compound present in various plant species. It is an active component of Phyllanthus urinaria and Terminalia catappa L extract used to treat various ailments.

Application

Corilagin has been used to study its inhibitory effect on the growth of breast cancer by inducing apoptosis and autophagy regulated by the release of reactive oxygen species.

Biochem/physiol Actions

Corilagin is a polyphenol, specifically a hydrolyzable tannin, found in Traditional Chinese Medicine herbal preparations. It is reported to have anti-inflammatory and anti-tumor activities.
Corilagin shows a variety of biological activities such as anti-inflammatory, anti-viral, anti-oxidant, hepatoprotective, antibacterial, anti-coagulant, hypolipemic and hypotensive. It has been noted to possess anti-tumor, antiproliferative, apoptotic, and autophagic abilities as well. Corilagin has been found to reduce the minimum inhibitory concentration (MIC) of β-lactams against Methicillin-resistant Staphylococcus aureus (MRSA).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Shimizu et al.
Antimicrobial agents and chemotherapy, 45(11), 3198-3201 (2001-10-16)
We found that an extract of Arctostaphylos uva-ursi markedly reduced the MICs of beta-lactam antibiotics, such as oxacillin and cefmetazole, against methicillin-resistant Staphylococcus aureus. We isolated the effective compound and identified it as corilagin. Corilagin reduced the MICs of various
Desmond Kwok Po Hau et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 16(8), 751-760 (2009-04-24)
Acetaminophen is a commonly used drug for the treatment of patients with common cold and influenza. However, an overdose of acetaminophen may be fatal. In this study we investigated whether mice, administered intraperitoneally with a lethal dose of acetaminophen, when
Nuchanart Rangkadilok et al.
Journal of agricultural and food chemistry, 53(5), 1387-1392 (2005-03-03)
Regular consumption of fruit and vegetables is associated with a lower risk of some chronic diseases including various forms of cancer and cardiovascular diseases. The health-promoting potential of these foods may be due, in part, to the phytochemical bioactive compounds
Xiao-Rong Dong et al.
International journal of molecular medicine, 25(4), 531-536 (2010-03-04)
Microglia, the resident immune cells of the central nervous system (CNS), are activated by various stimuli. Resting microglia are the basis of normal neurogenesis, while activated microglia may inhibit neurogenesis through the production of pro-inflammatory mediators and cytokines. Recent research
Zhi-Qiang Shen et al.
Planta medica, 69(12), 1109-1112 (2004-01-30)
In this study, Charlton's and Tomihisa's methods were modified to investigate the thrombolytic effect of corilagin from the Chinese herbal plant Phyllanthus urinaria L., as well as its effect on carotid artery patency status. The activity of type 1 plasminogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service