All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H6O8 · xH2O
Molecular Weight:
302.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
grade
technical grade
Quality Level
form
powder
mp
>300 °C (lit.)
SMILES string
O.Oc1cc2C(=O)Oc3c(O)c(O)cc4C(=O)Oc(c1O)c2-c34
InChI
1S/C14H6O8.H2O/c15-5-1-3-7-8-4(14(20)22-11(7)9(5)17)2-6(16)10(18)12(8)21-13(3)19;/h1-2,15-18H;1H2
InChI key
UOZZJRXBNGVIKO-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nina A Hering et al.
Frontiers in pharmacology, 12, 610164-610164 (2021-03-30)
Scope: Ellagitannins are polyphenols found in numerous fruits, nuts and seeds. The elagitannin punicalagin and its bioactive metabolites ellagic acid and urolithins are discussed to comprise a high potential for therapeutically or preventive medical application such as in intestinal diseases.
Silviya Abarova et al.
Biochimica et biophysica acta, 1863(11), 2934-2941 (2017-08-06)
Differential scanning calorimetry was applied to evaluate the efficacy of preventive treatments with biologically active compounds of plant origin against neurodegenerative disorder in mice. As we reported recently, large differences exist between the heat capacity profiles of water-soluble brain proteome
Lyubka P Tancheva et al.
Antioxidants (Basel, Switzerland), 9(1) (2020-01-16)
We compared the neuroprotective action of three natural bio-antioxidants (AOs): ellagic acid (EA), α-lipoic acid (LA), and myrtenal (Myrt) in an experimental model of Parkinson's disease (PD) that was induced in male Wistar rats through an intrastriatal injection of 6-hydroxydopamine
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service