F8509
Fumaric acid
BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
Beilstein:
605763
EC Number:
MDL number:
UNSPSC Code:
12352207
eCl@ss:
39021709
PubChem Substance ID:
NACRES:
NA.71
Recommended Products
grade
reagent
vapor pressure
1.7 mmHg ( 165 °C)
product line
BioReagent
Assay
≥98%
form
powder
autoignition temp.
1364 °F
expl. lim.
40 %
technique(s)
cell culture | mammalian: suitable
mp
298-300 °C (subl.) (lit.)
SMILES string
OC(=O)\C=C\C(O)=O
InChI
1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+
InChI key
VZCYOOQTPOCHFL-OWOJBTEDSA-N
Looking for similar products? Visit Product Comparison Guide
Application
A TCA (Krebs cycle) molecule used as a supplement in insect and mammalian cell culture.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
523.4 °F
Flash Point(C)
273 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tahar Ghnaya et al.
Chemosphere, 90(4), 1449-1454 (2012-10-03)
The implication of organic acids in Pb translocation was studied in two species varying in shoot lead accumulation, Sesuvium portulacastrum and Brassica juncea. Citric, fumaric, malic and α-cetoglutaric acids were separated and determined by HPLC technique in shoots, roots and
Emine Akar et al.
Carbohydrate polymers, 90(4), 1634-1641 (2012-09-05)
A novel biodegradable sodium carboxymethyl cellulose (NaCMC)-based hydrogel was synthesized by using fumaric acid (FA) as a crosslinking agent at various ratios. Hydrogels (CMCF) were characterized using Fourier transform infrared spectroscopy (FTIR), thermogravimetric analysis (TGA), differential scanning calorimetry (DSC), X-ray
Bas J Meussen et al.
Applied microbiology and biotechnology, 94(4), 875-886 (2012-04-25)
Rhizopus oryzae is a filamentous fungus belonging to the Zygomycetes. It is among others known for its ability to produce the sustainable platform chemicals L: -(+)-lactic acid, fumaric acid, and ethanol. During glycolysis, all fermentable carbon sources are metabolized to
Carol A Roa Engel et al.
Applied microbiology and biotechnology, 78(3), 379-389 (2008-01-25)
The potential of fumaric acid as a raw material in the polymer industry and the increment of cost of petroleum-based fumaric acid raises interest in fermentation processes for production of this compound from renewable resources. Although the chemical process yields
Rohan A Davis et al.
Bioorganic & medicinal chemistry, 18(1), 14-18 (2009-12-08)
We have investigated the enzyme inhibition characteristics of a natural product (NP)-based phenolic library against a panel of human carbonic anhydrases (hCAs, EC 4.2.1.1) which included hCAs I and II (cytosolic) and hCA VA/VB (mitochondrial isoforms). Most of these compounds
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service