EP5495404007
Eppendorf® F-32x0.2 rotor
for 32 x 0.2-mL PCR tubes tubes or 4 x 8 PCR strips, incl. rotor cover, Centrifuge 5420
All Photos(1)
About This Item
UNSPSC Code:
41103900
NACRES:
NB.82
Recommended Products
feature
for 32 x 0.2-mL PCR tubes tubes or 4 x 8 PCR strips, incl. rotor cover, Centrifuge 5420
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 5495404007
parameter
10,000 rpm speed
storage temp.
room temp
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)