EP5490040007
Rotor for Eppendorf® Concentrator Plus
F-40-36-12, non aerosol tight, to hold, 36 x 1.5 mL flat-bottom tubes
All Photos(1)
About This Item
Recommended Products
feature
to hold 36 x 1.5 mL flat-bottom tubes
to hold 36 x 12 x 32 mm
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490040007 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
General description
Selection of rotors for the Eppendorf Concentrator Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG