Key Documents
EP4960800155
Eppendorf® Bottle Thread Adapter
from GL 45 to GL 38, polypropylene
About This Item
UNSPSC Code:
41121815
NACRES:
NB.17
Pricing and availability is not currently available.
material
polypropylene
feature
from GL 45 to GL 38
manufacturer/tradename
Eppendorf® 4960800155
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
EP4960800120EP4960800066EP4960800040
manufacturer/tradename
Eppendorf® 4960800155
manufacturer/tradename
Eppendorf® 4960800120
manufacturer/tradename
Eppendorf® 4960800066
manufacturer/tradename
Eppendorf® 4960800040
feature
from GL 45 to GL 38
feature
from GL 45 to GL 32
feature
from GL 32 to GL 38
feature
from GL 32 to GL 25
General description
Eppendorf dispensers are designed for bottle threads of 45 mm. For other bottle threads a thread adapter can be used. The diameter of the thread adapter is imprinted on the adapter. If the bottle thread is smaller than 32 mm, a second adapter must be used. If you need a thread adapter with a higer resistance to chemicals, use a PTFE/ETFE adapter.
Application
Connection of Eppendorf dispensers to bottles with neck threads different to 45 mm.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG