Z742267
DURAN® GL45 thread adapter for bottle top vacuum filtration unit
GL 45 male, GL 56 female, white PTFE
Synonym(s):
291195601
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
white PTFE
manufacturer/tradename
DURAN® 291195601
fitting
GL 45 male
GL 56 female
Related Categories
General description
The re-usable adaptor (GL 45 external/GL 65) allows the use of the DURAN TILT bottle with the 45 mm filtration units for the fitler and sterilization of cell culture media. Manufactured from inert PTFE, can be autoclaved and depyrogenised at 300°C.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)