Skip to Content
Merck
All Photos(1)

Key Documents

EHU107101

Sigma-Aldrich

MISSION® esiRNA

targeting human PPIA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGTGTTTGGCAAAGTGAAAGAAGGCATGAATATTGTGGAGGCCATGGAGCGCTTTGGGTCCAGGAATGGCAAGACCAGCAAGAAGATCACCATTGCTGACTGTGGACAACTCGAATAAGTTTGACTTGTGTTTTATCTTAACCACCAGATCATTCCTTCTGTAGCTCAGGAGAGCACCCCTCCACCCCATTTGCTCGCAGTATCCTAGAATCTTTGTGCTCTCGCTGCAGTTCCCTTTGGGTTCCATGTTTTCCTTGTTCCCTCCCATGCCTAGCTGGATTGCAGAGTTAAGTTTATGATTATGAAATAAAAACTAAATAACAATTGTCCTCGTTTGAGTTAAGAGTGTTGATGTAGGCTTTATTTTAAGCAGTAATGGGTTACTTCTGAAACATCACTTGTTTGCTTAATTCTACACAGTACTTAGATTTTTTTTACTTTCCAGTCCCAGGAAGTGTCAATGTTTGTTGAGTGGAATATTGAAAATGTAGGCAGCAACTGGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huan Zhang et al.
PloS one, 9(3), e92824-e92824 (2014-03-26)
Hypoxia-inducible factor-1α (HIF-1α) is a highly important transcription factor involved in cell metabolism. HIF-1α promotes glycolysis and inhibits of mitochondrial respiration in pancreatic ductal adenocarcinoma (PDAC). In response to tumor hypoxia, cyclophilin A (CypA) is over-expressed in various cancer types
Hiroya Fujioka et al.
Oncology letters, 13(1), 289-295 (2017-01-27)
Paclitaxel is widely used to treat various cancers; however, resistance to this drug is a major obstacle to breast cancer chemotherapy. To identify the proteins involved in paclitaxel resistance, the present study compared the proteomes of MCF-7 human breast cancer
N Doti et al.
Cell death & disease, 5, e993-e993 (2014-01-18)
Delayed neuronal cell death largely contributes to the progressive infarct development and associated functional impairments after cerebral ischemia or brain trauma. Previous studies exposed a key role for the interaction of the mitochondrial protein apoptosis-inducing factor (AIF) and cytosolic cyclophilin
Lin Cong et al.
BioMed research international, 2020, 9210891-9210891 (2020-03-19)
In human pancreatic ductal adenocarcinoma (PDAC), the cyclophilin A (CypA) is overexpressed and promotes the development of PDAC. However, the mechanism underlying cyclophilin A expression remains elusive. Here, we reported that the citron Rho-interacting serine/threonine kinase (CIT) promotes the HIF1a-CypA
Hiroaki Takeuchi et al.
Retrovirology, 9, 3-3 (2012-01-10)
An understanding of host cell factors that affect viral replication contributes to elucidation of the mechanism for determination of viral tropism. Cyclophilin A (CypA), a peptidyl-prolyl cis-trans isomerase (PPIase), is a host factor essential for efficient replication of human immunodeficiency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service