Skip to Content
Merck
All Photos(1)

Key Documents

D0697

Sigma-Aldrich

DMEM/F-12

StableCell, with stable glutamine, HEPES and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, DME/F-12, 1:1 mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

StableCell DMEM/F12, With stable glutamine, 15mM HEPES and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

glucose: high
sodium pyruvate: yes
stable glutamine: yes
NaHCO3: yes
phenol red: yes
HEPES: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This DMEM F-12 Ham medium differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.

Legal Information

StableCell is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Jin et al.
Frontiers in molecular biosciences, 8, 717771-717771 (2021-10-16)
Airway smooth muscle cells (ASMCs) exist in a form of helical winding bundles within the bronchial airway wall. Such tubular tissue provides cells with considerable curvature as a physical constraint, which is widely thought as an important determinant of cell
Vivek Pandey et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 131, 110751-110751 (2020-11-07)
Deoxyelephantopin (DOE), a phytochemical, extracted and purified from Elephantopus scaber, has been shown to exhibit antitumor activities. Objective of the present study was to investigate anti-tumor and apoptosis-inducing properties of DOE against uterine leiomyoma (UL) and to explore their molecular
Fei Leng et al.
Oncology letters, 22(4), 697-697 (2021-08-31)
HOXB-AS3 is a long non-coding RNA and recent studies have shown that the HOXB-AS3-encoded micro-peptide was associated with the progression of colon cancer tumorigenesis; however, the biofunction of HOXB-AS3 varies in different types of cancer and the potential function in
Irene Cervelló et al.
PloS one, 6(6), e21221-e21221 (2011-06-30)
Endometrial regeneration is mediated, at least in part, by the existence of a specialized somatic stem cell (SSC) population recently identified by several groups using the side population (SP) technique. We previously demonstrated that endometrial SP displays genotypic, phenotypic and
Hitomi Nomura et al.
BMC cancer, 8, 39-39 (2008-02-02)
Gelsolin-like actin-capping protein (CapG) is a ubiquitous gelsolin-family actin-modulating protein involved in cell signalling, receptor-mediated membrane ruffling, phagocytosis, and motility. CapG has generated great interest due to its oncogenic function in the control of cell migration or invasion in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service