Skip to Content
Merck
All Photos(1)

Key Documents

EHU008611

Sigma-Aldrich

MISSION® esiRNA

targeting human IFIH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGCATGGAGGAGGAACTGTTGACAATTGAAGACAGAAACCGGATTGCTGCTGCAGAAAACAATGGAAATGAATCAGGTGTAAGAGAGCTACTAAAAAGGATTGTGCAGAAAGAAAACTGGTTCTCTGCATTTCTGAATGTTCTTCGTCAAACAGGAAACAATGAACTTGTCCAAGAGTTAACAGGCTCTGATTGCTCAGAAAGCAATGCAGAGATTGAGAATTTATCACAAGTTGATGGTCCTCAAGTGGAAGAGCAACTTCTTTCAACCACAGTTCAGCCAAATCTGGAGAAGGAGGTCTGGGGCATGGAGAATAACTCATCAGAATCATCTTTTGCAGATTCTTCTGTAGTTTCAGAATCAGACACAAGTTTGGCAGAAGGAAGTGTCAGCTGCTTAGATGAAAGTCTTGGACATAACAGCAACATGGGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Lin Chen et al.
Nature communications, 8, 13882-13882 (2017-01-10)
B-cell infection by hepatitis C virus (HCV) has been a controversial topic. To examine whether HCV has a genetically determined lymphotropism through a co-receptor specific for the infection by lymphotropic HCV, we established an infectious clone and chimeric virus of
Seung Bum Park et al.
PloS one, 11(7), e0158419-e0158419 (2016-07-13)
Hepatitis C virus (HCV) actively evades host interferon (IFN) responses but the mechanisms of how it does so are not completely understood. In this study, we present evidence for an HCV factor that contributes to the suppression of retinoic-acid-inducible gene-I
Ian T Lamborn et al.
The Journal of experimental medicine, 214(7), 1949-1972 (2017-06-14)
MDA5 is a cytosolic sensor of double-stranded RNA (ds)RNA including viral byproducts and intermediates. We studied a child with life-threatening, recurrent respiratory tract infections, caused by viruses including human rhinovirus (HRV), influenza virus, and respiratory syncytial virus (RSV). We identified
Tatsuro Saruga et al.
Molecular biology reports, 48(1), 425-433 (2021-01-03)
C-X-C motif chemokine 10 (CXCL10) is an inflammatory chemokine and a key molecule in the pathogenesis of rheumatoid arthritis (RA). Melanoma differentiation-associated gene 5 (MDA5) is an RNA helicase that plays a role in innate immune and inflammatory reactions. The
Tongtian Zhuang et al.
Cell death & disease, 11(6), 453-453 (2020-06-14)
Vitiligo is a disfiguring disease featuring chemokines-mediated cutaneous infiltration of autoreactive CD8+ T cells that kill melanocytes. Copious studies have indicated that virus invasion participates in the pathogenesis of vitiligo. IFIH1, encoding MDA5 which is an intracellular virus sensor, has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service