Skip to Content
Merck
All Photos(1)

Key Documents

E2777

Sigma-Aldrich

Monoclonal Anti-c-erbB-2 antibody produced in mouse

~2 mg/mL, clone HER2-96, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HER-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HER2-96, monoclonal

form

buffered aqueous solution

mol wt

antigen 185 kDa (complete protein)
antigen 97-115 kDa (extracellular ligand-binding domain)

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): 0.5-1 μg/test using RIPA lysate of cultured CB2 cells (CHO cell line transfected with erbB-2)
indirect ELISA: suitable
microarray: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

General description

Monoclonal Anti-c-erbB-2 (HER-2, Neu) (mouse IgG1 isotype) is derived from the (human epidermal growth factor receptor 2) HER2-96 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with a purified extracellular domain of human erbB-2, from transfected cells. The human erbB-2 oncogen (also known as HER2, HER-2 or neu) encodes a transmembrane growth factor orphan receptor with a M.W. of 185 kDa (p185). The complete protein consists of an internal cytoplasmic structure with tyrosine kinase activity, a short hydrophobic transmembrane section and an extracellular ligand-binding domain (ECD).

Immunogen

purified extracellular domain of human erbB-2 from transfected cells.

Application

Monoclonal Anti-c-erbB-2 antibody produced in mouse has been used in :
  • enzyme linked immunosorbent assay (ELISA)
  • immunoprecipitation
  • immunohistochemistry
  • studies on receptor internalization, phosphorylation, ligand binding and tumor growth inhibition

Biochem/physiol Actions

Erb-b2 receptor tyrosine kinase 2 (ErbB-2) has a pivotal role in signal transduction, where it can stimulate the signal even in the absence of ligand by forming heterodimers with erbB-1/EGF-R, erbB-3, and erbB-4 and hence can modifies ligand affinity. Thus, erbB-2 alters the intracellular responses elicited by EGF and NDF. This control is due to the fact that erbB-2, when in complex with another erbB family receptor, decelerates the rate of ligand dissociation. Therefore, erbB-2 may act as a signalling subunit for other receptors rather than a true growth factor receptor. Because overexpression of the erbB-2 proto-oncogene is frequently associated with an aggressive clinical course of certain human adenocarcinomas, the encoded protein is an attractive target for immunotherapy. Indeed, antibodies to erbB-2 were found to block the crosstalk with growth factor receptors. In addition, receptor overexpression is considered a predictor of poor survival and short time of relapse.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jekabs Krigerts et al.
Biophysical journal, 120(4), 711-724 (2021-01-17)
Finding out how cells prepare for fate change during differentiation commitment was our task. To address whether the constitutive pericentromere-associated domains (PADs) may be involved, we used a model system with known transcriptome data, MCF-7 breast cancer cells treated with
L N Klapper et al.
Oncogene, 14(17), 2099-2109 (1997-05-01)
ErbB-2 is an orphan receptor that belongs to a family of tyrosine kinase receptors for either epidermal growth factor (EGF) or Neu differentiation factor (NDF/neuregulin). Because overexpression of the erbB-2 proto-oncogene is frequently associated with an aggressive clinical course of
HER2-Associated Radioresistance of Breast Cancer Stem Cells Isolated from HER2-Negative Breast Cancer Cells
Duru A, et al.
Clinical Cancer Research, 18(24), 6634-6647 (2012)
Götz Pilarczyk et al.
International journal of molecular sciences, 18(2) (2017-02-18)
In cancer, vulnerable breast epithelium malignance tendency correlates with number and activation of ErbB receptor tyrosine kinases. In the presented work, we observe ErbB receptors activated by irradiation-induced DNA injury or neuregulin- 1 β application, or alternatively, attenuated by a
R Goldman et al.
Biochemistry, 29(50), 11024-11028 (1990-12-18)
The erbB-1 and erbB-2 protooncogenes encode homologous membrane receptors that respectively bind epidermal growth factor (EGF) and a still incompletely characterized ligand. Binding of EGF to its receptor is known to increase tyrosine phosphorylation of the erbB-2/neu receptor in tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service