Skip to Content
Merck
All Photos(3)

Key Documents

06-562

Sigma-Aldrich

Anti-erbB-2/HER-2 Antibody

Upstate®, from rabbit

Synonym(s):

CD340 antigen, CD340 antigen, NEU proto-oncogene, Tyrosine kinase-type cell surface receptor HER2, c-erb B2/neu protein, c-erb B2/neu protein, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2 (neuro/glioblastoma derived oncogene homolog),

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

monkey, human, mouse

species reactivity (predicted by homology)

rat

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)
mouse ... Erbb2(13866)
rat ... Erbb2(24337)
rhesus monkey ... Erbb2(697573)

General description

The ErbB-2 proto-oncogene, also called Neu, EGFR-2 or HER-2, is a member of the transmembrane receptor tyrosine kinase family, which also includes EGF receptor and EGFR-3 (HER-3 or ErbB-3). Although, ErbB-2 does not have any known high-affinity ligands, its kinase activity can be activated without ligand by either overexpression or heteroassociation with other members of the ErbB family. Amplification of the ErbB-2 gene and overexpression of its product has been detected in almost 40% of primary human breast tumors, which correlates with poor prognosis in node positive breast cancer. ErbB-2 overexpression is also observed in ovarian, gastric, salivary and non-small cell lung carcinomas. Because of this, ErbB-2 is one of the major drug targets for breast cancer and other cancer treatments.

Specificity

Recognizes c-erbB-2 (185 kDa), but not c-erbB-1(EGF-R), c-erbB-3 or c-erbB-4.

Immunogen

A KLH-linked peptide (AENPEYLGLDVPV) corresponding to aa 1243-1255 from the human c-erbB-2/HER-2/neu oncoprotein.

Application

Anti-erbB-2/HER-2 Antibody is an antibody against erbB-2/HER-2 for use in IH, IP & WB.
Immunoprecipitation:
4 μg of a previous lot immunoprecipitated erbB-2/HER-2 from 600 μg MCF-7 RIPA lysate, and previously in A431, LNCaP, and c-erbB-2 transfected 3T3/NIH RIPA lysate.

Immunohistochemistry:
A previous lot of antibody has been reported by an independent laboratory, to detect erbB-2/HER-2 in frozen and formalin/paraffin sections of rat breast.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Routinely evaluated by Western Blot on HEK293 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected Her-2 on 10 μg of HEK293 lysates.

Target description

185 kDa

Linkage

Replaces: 04-1127

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in buffer containing 10 mM PBS, pH 7.4, 0.2% BSA and 0.09% sodium azide. Frozen solution.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 cells, HEK293, Jurkat, MCF7 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pathologic findings from the National Surgical Adjuvant Breast and Bowel Project: prognostic significance of erbB-2 protein overexpression in primary breast cancer.
Paik, S, et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 8, 103-112 (1990)
Introducing the cancer microenvironment section of Journal of Translational Medicine.
Fernando Vidal-Vanaclocha,Isaac P Witz
Journal of Translational Medicine null
Different mechanisms are implicated in ERBB2 gene overexpression in breast and in other cancers.
Vernimmen, D; Gueders, M; Pisvin, S; Delvenne, P; Winkler, R
British Journal of Cancer null
Increased expression of epidermal growth factor receptor induces sequestration of extracellular signal-related kinases and selective attenuation of specific epidermal growth factor-mediated signal transduction pathways.
Amyn A Habib, Soo Jin Chun, Benjamin G Neel, Timothy Vartanian
Molecular Cancer Research null
Concomitant targeting of EGF receptor, TGF-beta and SRC points to a novel therapeutic approach in pancreatic cancer.
Deharvengt, S; Marmarelis, M; Korc, M
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service