Skip to Content
Merck
All Photos(1)

Key Documents

E1768

Sigma-Aldrich

5,8,11,14-Eicosatetraynoic acid

≥97%

Synonym(s):

ETYA, Octadehydroarachidonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Beilstein:
1798411
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥97%

form

powder

mp

78-80 °C

solubility

ethanol: 50 mg/mL, clear, colorless to yellow

storage temp.

−20°C

SMILES string

CCCCCC#CCC#CCC#CCC#CCCCC(O)=O

InChI

1S/C20H24O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-5,8,11,14,17-19H2,1H3,(H,21,22)

InChI key

MGLDCXPLYOWQRP-UHFFFAOYSA-N

Gene Information

human ... ALOX15(246)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Eicosatetraynoic acid (ETYA) is a non-metabolizable analog of ω-6 arachidonic acid. ETYA is a strong activator of the human peroxisome proliferator-activated receptor α (PPARα). It acts as an inhibitor of lipoxygenases (LOX) and cyclooxygenases (COX).
Eicosatetraynoic acid is a lipoxygenase and cyclooxygenase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuhiro Tamura et al.
Vascular pharmacology, 44(6), 411-416 (2006-05-03)
To address the role of prostaglandin E2 (PGE2) in tube formation of endothelial cells and the relationships between the action of PGE2 and vascular endothelial growth factor (VEGF), cultured human umbilical vein endothelial cells (HUVECs) were used to evaluate tube
Marcelo D Carattino et al.
The Journal of biological chemistry, 278(38), 36202-36213 (2003-07-03)
Epithelial Na+ channels (ENaCs) are regulated by the phospholipase A2 (PLA2) product arachidonic acid. Pharmacological inhibition of PLA2 with aristolochic acid induced a significant increase in amiloride-sensitive currents in Xenopus oocytes expressing ENaC. Arachidonic acid or 5,8,11,14-eicosatetraynoic acid (ETYA), a
Parissa Taheri et al.
Journal of plant physiology, 167(3), 201-208 (2009-09-05)
Vitamins are plant growth regulators and activators of defense responses against pathogens. The cytomolecular mechanisms involved in the induction of resistance by chemicals especially vitamins on monocotyledonous plants are largely unknown. Here, we show that riboflavin, which acts as a
Miriam Guizy et al.
American journal of physiology. Cell physiology, 289(5), C1251-C1260 (2005-07-01)
Dietary polyunsaturated fatty acids (PUFAs) have been reported to exhibit antiarrhythmic properties, which have been attributed to their availability to modulate Na(+), Ca(2+), and several K(+) channels. However, their effects on human ether-a-go-go-related gene (HERG) channels are unknown. In this
A S Taylor et al.
Prostaglandins, 29(3), 449-458 (1985-03-01)
5,8,11,14-eicosatetraynoic acid (ETYA), a widely used inhibitor of cyclooxygenase and lipoxygenase, inhibited the incorporation of 14C-arachidonic acid into cell lipids of the murine thymoma EL4 whereas oleic acid had no effect. Inhibition appeared to result from the ability of ETYA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service