Skip to Content
Merck
All Photos(2)

Key Documents

E3127

Sigma-Aldrich

cis-11,14-Eicosadienoic acid

≥98%, liquid

Synonym(s):

11,14-Eicosadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4(CH=CHCH2)2(CH2)8CO2H
CAS Number:
Molecular Weight:
308.50
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Quality Level

Assay

≥98%

form

liquid

bp

198 °C/0.08 mmHg (lit.)

application(s)

food and beverages

functional group

carboxylic acid

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCCCC(O)=O

InChI

1S/C20H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10H,2-5,8,11-19H2,1H3,(H,21,22)/b7-6-,10-9-

InChI key

XSXIVVZCUAHUJO-HZJYTTRNSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woo Jung Park et al.
Journal of lipid research, 50(6), 1195-1202 (2009-02-10)
The mammalian Delta6-desaturase coded by fatty acid desaturase 2 (FADS2; HSA11q12-q13.1) catalyzes the first and rate-limiting step for the biosynthesis of long-chain polyunsaturated fatty acids. FADS2 is known to act on at least five substrates, and we hypothesized that the
M J Steinbeck et al.
Journal of leukocyte biology, 49(4), 360-368 (1991-04-01)
To investigate NADPH-oxidase activation, we studied the effects of free fatty acid (FFA), their uncharged derivatives, and calcium on membrane lipid structure and superoxide anion (O2-) release from intact neutrophils and in cell-free O2(-)-generating systems. This study determined that in
Yung-Sheng Huang et al.
Molecular and cellular biochemistry, 358(1-2), 85-94 (2011-06-21)
Eicosadienoic acid (Δ11,14-20:2; EDA) is a rare, naturally occurring n-6 polyunsaturated fatty acid (PUFA) found mainly in animal tissues. EDA is elongated from linoleic acid (LA), and can also be metabolized to dihomo-γ-linolenic acid (DGLA), arachidonic acid (AA), and sciadonic
R Mogul et al.
Biochemistry, 40(14), 4391-4397 (2001-04-04)
Lipoxygenases are currently potential targets for therapies against asthma, artherosceloris, and cancer. Recently, inhibition studies on both soybean (SLO) and human lipoxygenase (15-HLO) revealed the presence of an allosteric site that binds both substrate, linoleic acid, and inhibitors; oleic acid
Y Angela Liou et al.
Prostaglandins, leukotrienes, and essential fatty acids, 80(4), 201-206 (2009-04-10)
High intakes of linoleic acid (LA,18:2n-6) have raised concern due to possible increase in arachidonic acid (ARA, 20:4n-6) synthesis, and inhibition of alpha linolenic acid (ALA, 18:3n-3) desaturation to eicosapentaenoic (EPA, 20:5n-3) and docosahexaenoic acid (DHA, 22:6n-3). In healthy men

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service