E0656
ER-27319
≥98% (HPLC), solid
Synonym(s):
10-(3-Aminopropyl)-3,4-dimethyl-9-acridone oxalate
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
solubility
DMSO: 28 mg/mL
storage temp.
2-8°C
SMILES string
OC(=O)C(O)=O.Cc1ccc2C(=O)c3ccccc3N(CCCN)c2c1C
InChI
1S/C18H20N2O.C2H2O4/c1-12-8-9-15-17(13(12)2)20(11-5-10-19)16-7-4-3-6-14(16)18(15)21;3-1(4)2(5)6/h3-4,6-9H,5,10-11,19H2,1-2H3;(H,3,4)(H,5,6)
InChI key
IIELZHYJYZBSGG-UHFFFAOYSA-N
1 of 4
This Item | A9789 | S5192 | D6317 |
---|---|---|---|
solid | solid | solid | solid |
DMSO: 28 mg/mL | H2O: 2 mg/mL (Aqueous solutions are stable over a wide range of pH at room temperature.), DMSO: 20 mg/mL, methanol: 20 mg/mL | DMSO: soluble ~28 mg/mL, H2O: insoluble | DMSO: ~28 mg/mL, H2O: insoluble |
2-8°C | 2-8°C | 2-8°C | 2-8°C |
Biochem/physiol Actions
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover Bioactive Small Molecules for Kinase Phosphatase Biology
Discover Bioactive Small Molecules for Kinase Phosphatase Biology
Discover Bioactive Small Molecules for Kinase Phosphatase Biology
Discover Bioactive Small Molecules for Kinase Phosphatase Biology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service