Skip to Content
Merck
All Photos(2)

Key Documents

D5294

Sigma-Aldrich

Dilazep dihydrochloride

powder

Synonym(s):

ASTA C 4898, Cormelian, N,N′-bis(3-[3,4,5-Trimethoxybenzoyloxy]propyl)homopiperazine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H44N2O10 · 2HCl
CAS Number:
Molecular Weight:
677.61
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

color

white

solubility

H2O: 10 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].COc1cc(cc(OC)c1OC)C(=O)OCCCN2CCCN(CCCOC(=O)c3cc(OC)c(OC)c(OC)c3)CC2

InChI

1S/C31H44N2O10.2ClH/c1-36-24-18-22(19-25(37-2)28(24)40-5)30(34)42-16-8-12-32-10-7-11-33(15-14-32)13-9-17-43-31(35)23-20-26(38-3)29(41-6)27(21-23)39-4;;/h18-21H,7-17H2,1-6H3;2*1H

InChI key

VILIWRRWAWKXRW-UHFFFAOYSA-N

General description

Dilazep comprises central cyclic diamine and phenyl rings connected via alkyl linkers.

Application

Dilazep dihydrochloride has been used in estrogen receptor (ERα) antagonistic assay. It has also been used as an equilibrative nucleoside transporter inhibitor (ENT 1 and 2) in cancer cell lines.

Biochem/physiol Actions

Dilazep is a equilibrative nucleoside transporter 1 (ENTs) inhibitor. It is an antianginal drug and an effective coronary vasodilator. Dilazep elicits anti-albuminuric effects and is an anti-platelet agent. It is a potent adenosine uptake inhibitor and suppresses effects of ischemia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Delaney et al.
The Journal of pharmacology and experimental therapeutics, 285(2), 568-572 (1998-05-15)
Selective inhibitors of adenosine production, degradation and transport were used to potentiate in vivo levels of adenosine and to determine the source of both basal and N-methyl-D-aspartate (NMDA)-induced increases in levels of endogenous adenosine in vivo. Male Sprague-Dawley rats receiving
H Deguchi et al.
Blood, 90(6), 2345-2356 (1997-10-06)
Dilazep, an antiplatelet agent, is generally used as an antithrombotic drug in clinical practice. Dilazep is also known to exert cytoprotective and antioxidant effects on endothelial cells. However, its effect on the endothelial or monocyte procoagulant activity is unknown. In
Dilazep analogues for the study of equilibrative nucleoside transporters 1 and 2 (ENT1 and ENT2)
Playa H, et al.
Bioorganic & medicinal chemistry letters, 24(24), 5801-5804 (2014)
Effects of dilazep on coronary and systemic hemodynamics in humans
Marzilli M, et al.
American Heart Journal, 108(2), 276-285 (1984)
Daniel Krys et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(12), 13837-13851 (2019-10-12)
Elevated proliferation rates in cancer can be visualized with positron emission tomography (PET) using 3'-deoxy-3'-l-[18F]fluorothymidine ([18F]FLT). This study investigates whether [18F]FLT transport proteins are regulated through hypoxia. Expression and function of human equilibrative nucleoside transporter (hENT)-1, hENT2, and thymidine kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service