Skip to Content
Merck
All Photos(1)

Key Documents

D-015

Supelco

Diphenhydramine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22ClNO
CAS Number:
Molecular Weight:
291.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2.Cl

InChI

1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H

InChI key

PCHPORCSPXIHLZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

Diphenhydramine is an antihistamine sold over the counter under the trade name Benadryl® and in other over the counter medications such as Tylenol PM, Advil PM, and Excedrin PM. Dihydramine is also consumed recreationally as a euphoriant and a deliriant.

Application


  • Hepatoprotective effects of polyphenols: Studies on the hepatoprotective effects of polyphenol-enriched fractions against oxidative stress in Acetaminophen-induced liver injury incorporate the analysis of Acetaminophen Related Compound D. This research underscores its relevance in pharmacology and toxicology, providing a deeper understanding of drug-induced liver injuries and potential therapeutic approaches (Wu et al., 2017).


Legal Information

Benadryl is a registered trademark of Johnson & Johnson
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Don W Coulter et al.
Anti-cancer drugs, 24(4), 415-421 (2013-01-19)
A pediatric study has established a maximum tolerated dose (MTD) for temsirolimus (Tem) of more than 150 mg/m intravenously/week. A phase I trial was conducted to establish the MTD for Tem in combination with valproic acid (VPA) in children and
Chang-Ik Choi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 909, 65-69 (2012-11-17)
Tamsulosin, a selective α₁-adrenoceptor antagonist, is used for the treatment of benign prostatic hyperplasia (BPH). We developed and validated a rapid, sensitive, and simplified liquid chromatography analytical method utilizing tandem mass spectrometry (LC-MS/MS) for the determination of tamsulosin in human
Benjamin W Friedman et al.
Annals of emergency medicine, 62(4), 311-318 (2013-04-10)
We compare metoclopramide 20 mg intravenously, combined with diphenhydramine 25 mg intravenously, with ketorolac 30 mg intravenously in adults with tension-type headache and all nonmigraine, noncluster recurrent headaches. In this emergency department (ED)-based randomized, double-blind study, we enrolled adults with
Marc Marino et al.
Journal of neuroscience methods, 211(1), 1-10 (2012-09-14)
Pruritus, the sensation of itch, which evokes reflex scratching behavior, has a diverse etiology. Because of its clinical significance, mechanisms of pruriception are an important topic. In the present work we describe and validate a paw motion detector (PMD) system.
A D Rollstin et al.
Clinical toxicology (Philadelphia, Pa.), 51(1), 50-53 (2012-12-01)
There are few reports of acetaminophen overdose in hypothermic patients and even fewer reports describing profound hypothermia. The kinetics, risk of hepatotoxicity, and the possible dose adjustments to N-acetylcysteine (NAC) therapy are not known in this setting. A 37-year-old female

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service