Skip to Content
Merck
All Photos(3)

Key Documents

D2636

Sigma-Aldrich

Dipalmitin

≥99.0%

Synonym(s):

Glycerol 1,2(3)-dihexadecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H68O5
CAS Number:
Molecular Weight:
568.91
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥99.0%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCCCCCC.CCCCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/2C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-31-33(36)32-40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-32-33(31-36)40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h2*33,36H,3-32H2,1-2H3

InChI key

UJTQAHPLAOYWPB-UHFFFAOYSA-N

Other Notes

Approx. 50% 1,2- and 50% 1,3-isomer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Aoyama et al.
The Journal of nutrition, 126(1), 225-231 (1996-01-01)
This study determined the effect of dietary (dipalmitoyl) triacylglycerol (TAG) structure on the absorption and metabolism of fat in the presence or absence of cholesterol. Sprague-Dawley male rats weighing approximately 180 g were fed diets containing 10 g/100 g diet
H Aubry et al.
Experimental biology, 47(2), 111-115 (1987-01-01)
The uptake of [H3] dipalmitoyl phosphatidylcholine by isolated brush border membranes from rabbit small intestine was studied. The process was found to be stimulated by Ca2+ in a manner which was influenced by the protein -Ca2+ concentration ratio. At higher
A Ortiz et al.
Biochemistry, 27(25), 9030-9036 (1988-12-13)
Mixtures of 1,2-dipalmitoylglycerol (1,2-DPG), 1,2-dioleylglycerol (1,2-DOG), 1,2-dicapryloylglycerol (1,2-DCG), 1,3-dioleylglycerol (1,3-DOG), and 1,3-dicapryloylglycerol (1,3-DCG) with dipalmitoylphosphatidylcholine (DPPC) have been studied by means of differential scanning calorimetry (DSC) and fluorescence depolarization of the probe diphenylhexatriene (DPH). DSC measurements showed that the tested
K V Rao et al.
Journal of neurochemistry, 63(4), 1454-1459 (1994-10-01)
The effect of arachidonic acid (AA) on the activity of diacylglycerol (DG) kinase in neural membranes was investigated. When rat brain cortical membranes were incubated with 0.5 mM dipalmitin and [gamma-32P]ATP, formation of phosphatidic acid (PA) was observed. It was
F López-García et al.
Biochimica et biophysica acta, 1190(2), 264-272 (1994-03-23)
The interaction of 1,2-dipalmitoylglycerol (1,2-DPG) with dipalmitoylphosphatidylserine (DPPS) has been studied in the presence and in the absence of Ca2+ by using differential scanning calorimetry (DSC) and 31P-nuclear magnetic resonance (31P-NMR). In the absence of Ca2+, DSC showed that 1,2-DPG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service