Key Documents
CLS6636
Corning® Step-R Repeating Pipettor and Syringe Tips Starter Pack
Synonym(s):
repeater
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Corning 6636
General description
For a smooth start, the Corning Step-R Repeating Pipettor Starter Pack is packaged with the Step-R Repeating Pipettor, 2 syringe tip adapters (25 mL and 50 mL), and 24 non-sterile syringe tips samples (0.05 mL, 2.5 mL, 5 mL, 12.5 mL, 25 mL, and 50 mL).
  • Easy volume setting: Volume selection chart conveniently placed on the handle
  • Fast and precise multiple dispensing: Up to 48 pipetting doses without refilling the tip syringe
  • Maintenance-free
  • Meets ISO 8655 requirements
  • Delivered with QC certificate
Legal Information
Corning is a registered trademark of Corning, Inc.