CLS6633
Corning® Step-R Repeating Pipettor Syringe Tips
volume (12.5 mL), sterile
Synonym(s):
pipet tips
All Photos(1)
About This Item
Recommended Products
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Corning 6633
volume
(12.5 mL)
General description
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Legal Information
Corning is a registered trademark of Corning, Inc.