Skip to Content
Merck
All Photos(1)

Key Documents

CLS432011

Corning® CoolCell SV10 Freezing Container

(6 x 10 mL serum vials)

Synonym(s):

Corning Coolcell accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300
NACRES:
NB.12

material

foamed PE

feature

Holds 6 x 10 mL vials

packaging

pkg of 1

manufacturer/tradename

Corning 432011

technique(s)

cryopreservation: suitable

capacity

(6 x 10 mL serum vials)

Legal Information

CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service