CLS431684
Corning® Erlenmeyer flask, 5 L
baffled bottom, polycarbonate, sterile
Synonym(s):
Corning container
All Photos(1)
About This Item
Recommended Products
material
flat bottom flask
polycarbonate
polypropylene cap
vent cap
wide-mouth
wide-mouth neck
sterility
sterile
feature
cap
closure type Vented cap
packaging
pack of 1 ea
case of 4 ea
manufacturer/tradename
Corning 431684
capacity
5 L
Features and Benefits
- Corning® Erlenmeyer flasks are ideal for shaker culture applications, and storage.
- Baffled bottom flasks are designed to scale up suspension culture efficiently.
- Molded-in graduations for accurate measurements.
- A large vent cap allows gas exchange and reduces spilling.
- Highest sterility assurance level (SAL) of 10-6
- RNase-/DNase-free and nonpyrogenic.
- Flasks are autoclavable once at 121°C, 15psi for 20 minutes. Repeated autoclaving weakens the flask and is not recommended. Vent cap is not autoclavable.
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Corning is a registered trademark of Corning, Inc.