Skip to Content
Merck
All Photos(1)

Key Documents

CLS3887

Corning® 96 Well Half-Area Microplate

flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 25, sterile, lid: no

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

black polystyrene
flat bottom clear
round wells

sterility

sterile

feature

lid: no

packaging

bag of 25
pkg of 25 inner package

manufacturer/tradename

Corning 3887A

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

black

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service