Skip to Content
Merck
All Photos(1)

Key Documents

CLS3882

Corning® 96 Well Half-Area Microplate

flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black plate
black polystyrene
clear bottom
flat bottom clear
size:
96 wells
well D:
10.7 mm
well volume:
190 μL
well working volume:
25-125 μL
Pricing and availability is not currently available.

material

black plate
black polystyrene
clear bottom
flat bottom clear

sterility

sterile

feature

lid
skirt

packaging

bag of 20
pkg of 20 inner package

manufacturer/tradename

Corning 3882

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

clear

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate have flat bottoms with 190 μL total volume.
  • Bottoms are 60% thinner than conventional polystyrene plates.
  • Black polystyrene, matrix active group TC-treated.
  • Low background fluorescence.
  • Enabling readings down to 340nm.
  • Opaque walls to prevent well-to-well crosstalk.
  • Direct microscopic viewing with optically clear flat well bottom.
  • Applicable for top and bottom reading instruments.
  • Nonreversible lids with condensation ring to reduce contamination.
  • Sterilized by gamma radiation and nonpyrogenic.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service