Skip to Content
Merck
All Photos(1)

Key Documents

CLLS1098

Sigma-Aldrich

SW48 Cells HIF1A -/-/-

Human female colon, colorectal adenocarcinoma

Synonym(s):

CLL1008

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

product name

SW48 Cells HIF1A -/-/-,

biological source

human female colon (Source disease: Colorectal adenocarcinoma)

Quality Level

storage temp.

−196°C

Gene Information

human ... HIF1A(3091)

General description

SW48 Cells HIF1A -/-/- are colorectal adenocarinoma, epithelial cells, from a human adult female, with a ZFN knock out modification.
This product corresponds to ATCC Cat. No. CCL-231.

Application

Media Renenewal changes one to two times per week.
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture ratio: approx. 1:2 to 1:6.
The base medium for this cell line is DMEM, Cat. No. D5671. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10%, L-glutamine, Catalog No. G7513, to a final concentration of 2 mM and sodium pyruvate, Cat. No. S8636, to a final concentration of 1 mM.
Cell freezing medium-DMSO 1X, Cat. No. C6164.

Features and Benefits

These SW48 cells are adherent, with a doubling time of approx. 35 hours.
Zinc finger nuclease (ZFN) knock out on chromosome 14q23.2

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • SW48 CELLS HIF1A -/-/- 1 vial


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service