Skip to Content
Merck
All Photos(1)

Key Documents

CLLS1127

Sigma-Aldrich

DLD1 Cells TP53 -/-

human male colon (Source Disease: Colorectal adenocarcinoma)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

product name

DLD1 Cells TP53 -/-,

biological source

human male colon (Source Disease: Colorectal adenocarcinoma)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... TP53(7157)

General description

DLD1 Cells TP53-/- are colorectal adenocarcinoma, epithelial cells, from a human adult male, with a ZFN double knock out modification.

This product corresponds to ATCC Cat. No. CCL-221.

Application

Media Renewal changes two to three times per week.

Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.

Subculture Ratio: approx. 1.3-1.10

The base medium for this cell line is RPMI, Catalog No. R5886. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Catalog No. F4135, to a final concentration (v/v) of 10%, L-glutamine, Catalog No. G7513, to a final concentration of 2 mM and sodium pyruvate, Catalog No. S8636, to a final concentration of 1 mM.

The cell freezing medium is DMSO 1X (Catalog Number C6164).

Features and Benefits

Zinc finger nuclease (ZFN) double knock out on chromosome 17p13.1

The DLD-1 cells are adherent, with a doubling time of approx. 20 hours.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, and PCR assay for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • DLD1 Cells TP53 -/- 1 vial

  • DLD1 Control Cells 1 vial


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service