Skip to Content
Merck
All Photos(1)

Key Documents

C9115

Sigma-Aldrich

CP-24879 hydrochloride

≥98% (HPLC), powder

Synonym(s):

p-Isopentoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18NOCl
CAS Number:
Molecular Weight:
215.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to brown

mp

154-159 °C

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

Cl[H].CC(C)CCOc1ccc(N)cc1

InChI

1S/C11H17NO.ClH/c1-9(2)7-8-13-11-5-3-10(12)4-6-11;/h3-6,9H,7-8,12H2,1-2H3;1H

InChI key

GFESZSNFRSACMU-UHFFFAOYSA-N

Biochem/physiol Actions

delta5/delta6 (Δ5/Δ6) desaturase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Yoon Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(51), 32433-32442 (2020-12-09)
Ferroptosis is an iron-dependent regulated necrosis mediated by lipid peroxidation. Cancer cells survive under metabolic stress conditions by altering lipid metabolism, which may alter their sensitivity to ferroptosis. However, the association between lipid metabolism and ferroptosis is not completely understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service