Skip to Content
Merck
All Photos(1)

Key Documents

PZ0176

Sigma-Aldrich

SC-26196

≥98% (HPLC)

Synonym(s):

SC-26196, a,a-diphenyl-4-[(3-pyridinylmethylene)amino]-1-piperazinepentanenitrile; 2,2-diphenyl-5-(4-[[(1 E)-pyridin-3-yl-methylidene]amino]piperazin-1-yl)pentanenitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29N5
CAS Number:
Molecular Weight:
423.55
MDL number:
UNSPSC Code:
41121801
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

N#CC(CCCN1CCN(CC1)\N=C\c2cccnc2)(c3ccccc3)c4ccccc4

InChI

1S/C27H29N5/c28-23-27(25-10-3-1-4-11-25,26-12-5-2-6-13-26)14-8-16-31-17-19-32(20-18-31)30-22-24-9-7-15-29-21-24/h1-7,9-13,15,21-22H,8,14,16-20H2/b30-22+

InChI key

QFYKXKMYVYOUNJ-JBASAIQMSA-N

Application

SC-26196 has been used as an inhibitor of delta6 (Δ6) fatty acid desaturase:
  • in mouse inner medullary collecting duct (IMCD3) and human (female) embryonic kidney (HEK) 293 cell culture as Dulbecco′s modified eagle′s medium (DMEM) component
  • in hepatic HepG2 cells
  • in glioblastoma cell lines to test its effect post-radiation treatments

Biochem/physiol Actions

SC-26196 is a Delta6 fatty acid desaturase (Delta6D) inhibitor. It specifically inhibited Delta6D activity with an IC(50) value of 100 nM. The rate-limiting step in arachidonic acid synthesis is the desaturation of dietary linoleic acid by Delta6D. SC-26196 completely prevented this conversion of linoleic acid to arachidonic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wondong Kim et al.
Cell metabolism, 29(4), 856-870 (2019-01-29)
The reactions catalyzed by the delta-5 and delta-6 desaturases (D5D/D6D), key enzymes responsible for highly unsaturated fatty acid (HUFA) synthesis, regenerate NAD+ from NADH. Here, we show that D5D/D6D provide a mechanism for glycolytic NAD+ recycling that permits ongoing glycolysis
Wan-Hsin Chang et al.
Lipids in health and disease, 17(1), 201-201 (2018-08-30)
The macrophage plays an important role in innate immunity to induce immune responses. Lipid replacement therapy has been shown to change the lipid compositions of mitochondria and potentially becomes an alternative to reduce the inflammatory response. We examined the effects
Jie Wang et al.
Cancer management and research, 10, 6779-6790 (2018-12-26)
It has been reported that cell inflammation pathways contribute to the development of prostaglandin E2 (PGE2)-inhibitor of DNA-binding protein-1 (ID1)-dependent radio-resistance in glioblastoma. Here, we proposed that inhibiting delta-6-desaturase (D6D) could block arachidonic acid synthesis and PGE2 production, thereby reversing
Aiguo Wu et al.
Biochimica et biophysica acta, 1852(5), 951-961 (2015-01-01)
Dietary deficiency of docosahexaenoic acid (C22:6 n-3; DHA) is linked to the neuropathology of several cognitive disorders, including anxiety. DHA, which is essential for brain development and protection, is primarily obtained through the diet or synthesized from dietary precursors, however
Jiajun Du et al.
Nature communications, 11(1), 4830-4830 (2020-09-26)
Non-invasively probing metabolites within single live cells is highly desired but challenging. Here we utilize Raman spectro-microscopy for spatial mapping of metabolites within single cells, with the specific goal of identifying druggable metabolic susceptibilities from a series of patient-derived melanoma

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service