Skip to Content
Merck
All Photos(1)

Key Documents

C7673

Sigma-Aldrich

Monoclonal Anti-CD14 antibody produced in mouse

clone UCHM-1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-CD14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

UCHM-1, monoclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

human

technique(s)

flow cytometry: 5 μL using 1 × 106 cells

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD14(929)

Looking for similar products? Visit Product Comparison Guide

Specificity

The epitope recognized by this clone is sensitive to routine formalin fixation and paraffin embedding. Cryostat sections, post-fixed in formalin can be stained.
3rd Workshop: code no. 203
4th Workshop: code no. M24

Immunogen

human thymocytes followed by peripheral blood T cells.

Application

Monoclonal Anti-CD14 antibody produced in mouse is suitable for the following applications:
  • Flow cytometry at a dilution of 5μL using 1 × 106 cells
  • Immunofluorescence.
  • Immunoblotting.

Biochem/physiol Actions

CD14 gene has an important role during signaling of the innate immune response and in the detection of inflammatory pathogens. It also plays an important role in recognizing tuberculosis (TB) disease and it can be used as a potential biomarker for predicting the prognosis of breast invasive ductal carcinoma with LN+ER/PR-Her2+. It is also associated with asthma, croup and allergy.

Target description

CD14 is a phosphoinositol-linked, single-chain, human cell surface glycoprotein antigen which is found on most human peripheral blood monocytes, granulocytes and dendritic reticulum cells, and some tissue macrophages. CD14 may also be present on B cells. Soluble forms of CD14 are present in serum and urine. The antigen functions as an acceptor for LPS-binding protein complexes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea D Lehmann et al.
Particle and fibre toxicology, 6, 26-26 (2009-10-10)
Using an in vitro triple cell co-culture model consisting of human epithelial cells (16HBE14o-), monocyte-derived macrophages and dendritic cells, it was recently demonstrated that macrophages and dendritic cells create a transepithelial network between the epithelial cells to capture antigens without
Weifeng He et al.
PloS one, 8(9), e75366-e75366 (2013-10-03)
In clinical practice, breast cancers with lymph node positive, ER/PR-negative and overexpressed human epidermal growth factor receptor 2 (LN+ER/PR-Her2+) have high risk of recurrence, but the effective biomarkers of prognostic for this type tumor are still lacking. Since breast cancers
Melissa K Callahan et al.
Journal of leukocyte biology, 74(5), 846-856 (2003-09-10)
Expression of phosphatidylserine (PS) on the surface of both macrophages and their apoptotic targets is required for efficient phagocytosis. Monocytes, the precursors of macrophages, do not express PS on their surface and do not efficiently phagocytose apoptotic cells. We report
Donna C Rennie et al.
Disease markers, 35(6), 765-771 (2013-12-19)
The CD14 gene has an important role in the detection of inflammatory provoking pathogens and in the ensuing signaling of the innate immune response. We assessed the role of CD14 C-159T, G-1359T in the expression of asthma, croup, and allergy
J Zhao et al.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease, 17(11), 1472-1478 (2013-10-16)
CD14 plays an important role in recognising the tuberculosis (TB) antigen and initiating immune response. CD14-159C/T polymorphism has been reported to be associated with susceptibility to TB in some, but not all studies. To comprehensively evaluate the correlation between CD14-159C/T

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service