Skip to Content
Merck
All Photos(1)

Key Documents

SRP3211

Sigma-Aldrich

IL-4 from mouse

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

B cell stimulating factor (BSF-1), BCDF, BCGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤2.0 ng/mL ED50

mol wt

13.7 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... IL4(16189)

General description

IL-4 (interleukin 4) is a pleiotropic cytokine. Produced by mast cells, T cells and bone marrow stromal cells. Recombinant murine IL-4 is a 13.7kDa globular protein containing 120 amino acid residues.

Biochem/physiol Actions

It regulates diverse T and B cell responses including cell proliferation, survival and gene expression. IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 (T helper) cells, characterized by their cytokine-secretion profile that includes secretion of IL-4, IL-5, IL-6, IL-10, and IL-13, which favor a humoral immune response. Another dominant function of IL-4 is the regulation of immunoglobulin class switching to the IgG1 (immunoglobulin G1) and IgE isotypes. Excessive IL-4 production by Th2 cells has been associated with elevated IgE production and allergy. IL-4 helps in long-term recovery after cerebral ischemia by controlling the activities of microglia/macrophage. In mice, absence of IL-4 is associated with a state anxiety-like behavior.

Sequence

MHIHGCDKNH LREIIGILNE VTGEGTPCTE MDVPNVLTAT KNTTESELVC RASKVLRIFY LKHGKTPCLK KNSSVLMELQ RLFRAFRCLD SSISCTMNES KSTSLKDFLE SLKSIMQMDY S

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Wei Ko et al.
Bio-protocol, 8(17), e3003-e3003 (2018-09-05)
Using gas chromatography mass spectrometry (GC-MS) to analyze the citric acid cycle (CAC) and related intermediates (such as glutamate, glutamine, GABA, and aspartate) is an analytical approach to identify unexpected correlations between apparently related and unrelated pathways of energy metabolism.
Mast cell production and response to IL-4 and IL-13.
McLeod JJ
Cytokine, 75, 57-61 (2015)
Interleukin-4 Is Essential for Microglia/Macrophage M2 Polarization and Long-Term Recovery After Cerebral Ischemia.
Liu X
Stroke, 47, 498-504 (2016)
Ile50Val variant of IL4R alpha upregulates IgE synthesis and associates with atopic asthma.
Mitsuyasu H
Nature Genetics, 19, 119-120 (1998)
Saeko Nakajima et al.
The Journal of investigative dermatology, 134(8), 2122-2130 (2014-02-01)
Atopic dermatitis (AD) is generally regarded as a type 2 helper T (Th2)-mediated inflammatory skin disease. Although the number of IL-17A-producing cells is increased in the peripheral blood and in acute skin lesion of AD patients, the role of IL-17A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service