Skip to Content
Merck
All Photos(1)

Key Documents

C5688

Sigma-Aldrich

Complement factor D from human plasma

90-110 μg/mL in PBS, pH 7.2, ≥90% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human plasma

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

concentration

90-110 μg/mL in PBS, pH 7.2

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CFD(1675)

Application

Complement factor D is a serine protease essential for the activation of the alternative pathway in the complement system. Research of complement factor D has revealed that a self inhibitory loop is responsible for switch from a resting to active state. Additionally, recent research has linked a complement factor D gene, single nucleotide polymorphism (SNP) rs3826945, with age-related macular degeneration (AMD). In these AMD cases there was a significant increase in plasma complement factor D concentration, even though previous research has shown no association between complement factor D SNP genes and AMD.

Biochem/physiol Actions

Complement factor D is a serine protease with structural homologies with chymotrypsin. In the complement cascade, factor D cleaves complement factor B in complex with activated complement component C3. It is identical to adipsin, which is expressed in adipose tissue.

Analysis Note

Source plasma tested for HBSAg, HCV, HBC, syphillis, and HIV-1/HIV-2

Other Notes

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

inhibitor

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M A Niemann et al.
Journal of immunology (Baltimore, Md. : 1950), 132(2), 809-815 (1984-02-01)
The monoclonal antibodies (MAb) were studied for their binding specificities and their effects on the hemolytic and proteolytic activities of human D. One Mab, FD10-1, was obtained from mice immunized with native D; the other, JA4-2, was similarly obtained from
Jiexi Zeng et al.
Molecular vision, 16, 2273-2278 (2010-12-09)
Age-related macular degeneration (AMD) is the most common cause of irreversible central vision loss worldwide. Research has linked AMD susceptibility with dysregulation of the complement cascade. Typically, complement factor H (CFH), complement factor B (CFB), complement component 2 (C2), and
Chloe M Stanton et al.
Investigative ophthalmology & visual science, 52(12), 8828-8834 (2011-10-18)
To examine the role of complement factor D (CFD) in age-related macular degeneration (AMD) by analysis of genetic association, copy number variation, and plasma CFD concentrations. Single nucleotide polymorphisms (SNPs) in the CFD gene were genotyped and the results analyzed
Shin Yong Park et al.
Journal of immunology (Baltimore, Md. : 1950), 181(9), 6328-6336 (2008-10-23)
Enterococcus faecalis (Ef) accounts for most cases of enterococcal bacteremia, which is one of the principal causes of nosocomial bloodstream infections (BSI). Among several virulence factors associated with the pathogenesis of Ef, an extracellular gelatinase (GelE) has been known to
H Jing et al.
Journal of molecular biology, 282(5), 1061-1081 (1998-10-01)
Factor D is a serine protease essential for the activation of the alternative pathway of complement. The structures of native factor D and a complex formed with isatoic anhydride inhibitor were determined at resolution of 2.3 and 1.5 A, respectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service