Skip to Content
Merck
All Photos(1)

Key Documents

C5572

Sigma-Aldrich

Click′s Medium

With sodium bicarbonate, without mercaptoethanol and L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Click’s EHAA Medium, Eagle’s Ham’s amino acids; EHAA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
phenol red: yes
glucose: yes

shipped in

ambient

storage temp.

2-8°C

Application

Click′s Medium has been used:
  • to incubate cartilage discs to measure the ability of chondrocytes to synthesize sulphated glycosaminoglycans (GAGs) by a metabolic assay
  • as a culture medium to incubate frozen peripheral blood mononuclear cells (PBMCs) in vitro stimulation and evaluation of immunologic response
  • for the suspension, washing and culture of T-cells

Reconstitution

Supplement with 0.584 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David E Pegg et al.
Cryobiology, 93, 12-17 (2020-04-25)
Although it is relatively straightforward to cryopreserve living isolated chondrocytes, at the present time there is no satisfactory method to preserve surgical grafts between the time of procurement or manufacture and actual use. In earlier papers we have established that
Expression of CD14, IL10, and Tolerogenic Signature in Dendritic Cells Inversely Correlate with Clinical and Immunologic Response to TARP Vaccination in Prostate Cancer Patients.
Luciano Castiello et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(13), 3352-3364 (2017-01-12)
David E Pegg et al.
Cryobiology, 52(3), 347-359 (2006-03-11)
Although isolated chondrocytes can be cryopreserved by standard methods, at the present time there is no satisfactory method that will preserve living chondrocytes in situ in surgical grafts, between the time of procurement or manufacture and actual use; survival of
Long Zheng et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(14), 3694-3706 (2020-04-11)
The murine Lym-1 mAb targets a discontinuous epitope (Lym-1 epitope) on several subtypes of HLA-DR, which is upregulated in a majority of human B-cell lymphomas and leukemias. Unlike CD19, the Lym-1 epitope does not downregulate upon crosslinking, which may provide

Related Content

Click's Medium Formulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service