Click's Medium Formulation
Component | C5572 [1x] g/L |
---|---|
Inorganic Salts | |
Calcium Chloride | 0.1854 |
Magnesium Chloride • 6H2O | 0.1649 |
Potassium Chloride | 0.4 |
Potassium Phosphate Dibasic | 0.06 |
Sodium Bicarbonate | 1.35 |
Sodium Chloride | 8 |
Sodium Phosphate Dibasic | 0.0479 |
Amino Acids | |
Alanine | 0.0356 |
Arginine • HCl | 0.3175 |
Asparagine • H2O | 0.06 |
L-Aspartic Acid | 0.0532 |
Cystine • 2HCl | 0.0782 |
Glutamic Acid | 0.0588 |
Glycine | 0.03 |
Histidine • HCl | 0.1047 |
Isoleucine | 0.13 |
Leucine | 0.13 |
L-Lysine • HCl | 0.1812 |
Methionine | 0.0375 |
Phenylalanine | 0.08 |
Proline | 0.046 |
Serine | 0.042 |
Threonine | 0.12 |
Tryptophan | 0.025 |
Tyrosine • 2Na | 0.1189 |
Valine | 0.115 |
Vitamins | |
D-Pantothenic Acid • ½Ca | 0.002 |
Choline Chloride | 0.002 |
Folic Acid | 0.002 |
myo-Inositol | 0.004 |
Niacinamide | 0.002 |
Pyridoxal • HCl | 0.002 |
Riboflavin | 0.0002 |
Thiamine • HCl | 0.002 |
Other | |
Glucose | 1 |
Phenol Red • Na | 0.01 |
Sodium Pyruvate | 0.275 |
Add | |
L-Glutamine | 0.584 |